Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

124014

Sigma-Aldrich

Akt Inhibitor VII, TAT-Akt-in

The Akt Inhibitor VII, TAT-Akt-in controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Akt Inhibitor VII, TAT-Akt-in, H- YGRKKRRQRRR-AVTDHPDRLWAWEKF-OH, TAT-TCL1₁₀₋₂₄, H-YGRKKRRQRRR-AVTDHPDRLWAWEKF-OH, TAT-TCL1₁₀₋₂₄

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C152H239N55O36
Molecular Weight:
3412.87
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable and reversible version of the Akt Inhibitor VI, Akt-in (Cat. No. 124013) fused with the protein transduction domain TAT that displays antitumor properties. Inhibits the phosphorylation of Akt selectively in HEK 293 and QRsP-11 fibrosarcoma cells stimulated with PDGF (complete inhibition at ~50 µM) with minimal inhibition towards PKA, PKC, PDK1, p42/44 MAPK, or p38 MAPK. Shown to prevent the proliferation of T4 cells, and tumor growth in syngeneic C57BL/6 mice.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: yes
Target IC50: 50 µM against Akt

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Sequence

H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Ala-Val-Thr-Asp-His-Pro-Asp-Arg-Leu-Trp-Ala-Trp-Glu-Lys-Phe-OH

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service