Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

12177

Millipore

Agar

Suitable for inclusion in culture media.

Synonym(s):

Agar-agar, Gum agar

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
MDL number:
UNSPSC Code:
50405901
NACRES:
NA.74
Pricing and availability is not currently available.

description

RTECS - AW7950000
Merck USA index - 14, 184

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

beige

solubility

hot water: soluble (Concentration is dependant on the % agar desired.)

shipped in

ambient

storage temp.

15-25°C

SMILES string

O1[C@@H]([C@@H]([C@@H]([C@H](C1OC2[C@H]3OCC2OC([C@H]3O)C)O)OC)O)CO

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Suitable for inclusion in culture media.

Application


  • Isolation of the Antifungal Compound Alliodorin from the Heartwood of Cordia elaeagnoides A. DC. and the In Silico Analysis of the Laccase.: The study isolates an antifungal compound from the heartwood of Cordia elaeagnoides, utilizing agar as a medium. The research highlights the potential of agar in facilitating the isolation and study of bioactive compounds in plant sciences (Guevara-Martínez et al., 2024).

  • Unveiling the Multifaceted Capabilities of Endophytic Aspergillus flavus Isolated from Annona squamosa Fruit Peels against Staphylococcus Isolates and HCoV 229E-In Vitro and In Silico Investigations.: This article examines the antimicrobial properties of Aspergillus flavus isolated using agar. The study reveals significant capabilities against Staphylococcus isolates and HCoV 229E, indicating agar′s critical role in microbiological research (Fathallah et al., 2024).

  • Phenotypic and Genomic Characterization of Pseudomonas wuhanensis sp. nov., a Novel Species with Promising Features as a Potential Plant Growth-Promoting and Biocontrol Agent.: The research characterizes a novel species of Pseudomonas, employing agar in the cultivation and analysis processes. The study underscores agar′s importance in microbial identification (Hou et al., 2024).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 1 month at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service