Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.19251

Supelco

Oxygen Scavengers Test, photometric

photometric, 0.09-2.17 mg/L (MEKO), 0.08-1.95 mg/L (ISA), 0.05-1.32 mg/L (Hydro), 0.027-0.667 mg/L (Carbohy), 0.020-0.500 mg/L (DEHA), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Oxygen Scavengers Test, photometric, 0.09-2.17 mg/L (MEKO), 0.08-1.95 mg/L (ISA), 0.05-1.32 mg/L (Hydro), 0.027-0.667 mg/L (Carbohy), 0.020-0.500 mg/L (DEHA), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 200 tests

analyte functional class(es)

oxygen scavengers

measuring range

0.020-0.500 mg/L (DEHA)
0.027-0.667 mg/L (Carbohy)
0.05-1.32 mg/L (Hydro)
0.08-1.95 mg/L (ISA)
0.09-2.17 mg/L (MEKO)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Iron reduction)

storage temp.

15-25°C

General description

Method: photometric 0.020 - 0.500 mg/l DEHA 0.027 - 0.667 mg/l Carbohy 0.053 - 1.315 mg/l Hydro 0.078 - 1.950 mg/l ISA 0.087 - 2.170 mg/l MEKO Spectroquant®

This Spectroquant® Oxygen Scavengers Reagent Test allows the accurate quantification of the oxygen scavengers content in aqueous samples.


Method applied: In the presence of oxygen scavengers iron(III) ions are reduced to iron(II) ions. These react with a triazine derivative (FerroZine) to form a violet complex which is determined photometrically.


The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.


All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Analysis Note

Performance test: conforms

Legal Information

FerroZine is a trademark of Hach Company
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service