Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.16801

Millipore

Agarose

(medium electroendoosmosis) for electrophoresis

Synonym(s):

Agarose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41105317
EC Index Number:
232-731-8
NACRES:
NB.21

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

mp

60-90 °C

bulk density

700 kg/m3

shipped in

ambient

storage temp.

−20°C

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

Warning

Toxicity: Standard Handling (A)

Analysis Note

Appearance (colour): white
Appearance (description): crystalline powder
Electroendoosmosis: 0.16 - 0.19
Sulphur (as SO₄): ≤ 0.2 %
Loss on drying (105 °C, 2 h): ≤ 8.0 %
Ash: ≤ 0.6 %
Gel point (1.5 % gel): 34 - 38 °C

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service