Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.15973

Sigma-Aldrich

Weigert′s iron hematoxylin kit

for nuclear staining in histology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.41

Quality Level

IVD

for in vitro diagnostic use

application(s)

clinical testing
diagnostic assay manufacturing
hematology

storage temp.

15-25°C

General description

Weigert′s iron hematoxylin kit - for nuclear staining in histology is a ready to use staining kit used for human-medical cell diagnosis and serves the purpose of the histological investigation of sample material of human origin, for example histological sections of e. g. the liver, the kidney, the intestine, the placenta and similar. Aluminium hematoxylins (e. g. nuclear fast red aluminium sulfate) are washed out of the tissue sections by acidic staining solutions. Iron hematoxylin solutions (e. g. Weigert′s iron hematoxylin), on the other hand, are acid-resistant and are preferentially used for staining nuclei in histological tissue sections in which counterstaining is done with acidic staining solutions. Examples here include the visualization of elastic fibers according to the van Gieson method (e. g. Elastica van Gieson (Product number 1.15974)) and the various trichromo connective-tissue staining methods (e. g. according to Masson-Goldner (Product number 1.00485). Commonly used hemalum solutions such as Mayer′s Hemalum, Delafield, Gill and Harris tend to procedure also weak nucleic staining only when used with acidic counter-stains.The kit contains two solutions; an alcoholic hematoxylin solution and a hydrochloric acid iron(III)nitrate solution, which are ready-to-use and should be applied at 3-5 μm paraffin sections.
The package is sufficient for 400 - 500 applications. Weigert′s Iron hematoxylin kit is an IVD & CE product and can be used in the clinical laboratory for diagnostics. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chi-Feng Weng et al.
Frontiers in physiology, 10, 1445-1445 (2020-01-11)
Adipose-derived stem cells (ADSCs) have shown great promise for the treatment of myocardial infarction (MI), although their potential therapeutic mechanism remains poorly understood. Growing evidence has implicated microRNAs (miRNAs or miRs) in the biological processes whereby ADSCs could ameliorate cardiovascular
C M Davies et al.
European cells & materials, 11, 57-75 (2006-04-14)
This paper introduces the culture preparation of ovine, bovine and human cancellous bone cores to be used in an explants model Zetos. The three dimensional (3D) bone cores were prepared and evaluated for all three animals. Bone cells in vivo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service