MilliporeSigma
  • Mechanically loaded ex vivo bone culture system 'Zetos': systems and culture preparation.
Mechanically loaded ex vivo bone culture system 'Zetos': systems and culture preparation.
European cells & materials (2006-04-14)
C M Davies, D B Jones, M J Stoddart, K Koller, E Smith, C W Archer, R G Richards
ABSTRACT
This paper introduces the culture preparation of ovine, bovine and human cancellous bone cores to be used in an explants model Zetos. The three dimensional (3D) bone cores were prepared and evaluated for all three animals. Bone cells in vivo constantly interact with each other, migratory cells, surrounding extracellular matrix (ECM) and interstitial fluid in a microenvironment, which continuously responds to various endogenous and exogenous stimuli. The Zetos system was designed to culture and mechanically load viable cancellous bone explants in their near natural microenvironment. This 3D ex vivo system bridges the current gap between in vitro and in vivo methods. One aim of this work was to compare the macro and micro-architecture of ovine, bovine and human cancellous bone tissue in preparation for culture within the Zetos system in order to determine the optimal source of experimental material. A second aim was to optimise the preparations of the bone cores as well as develop techniques involved during tissue maintenance. Bone core response was visualised using histological and immunohistochemical methods. The results demonstrate that cancellous bone explants vary greatly in trabecular density and bone volume depending on species, age and location. Sheep and human samples displayed the greatest variation between bones cores when compared to bovine. Even cores taken from the same animal possessed very different characteristics. The histology demonstrated normal bone and cell structure after the core preparation. Immunohistochemistry results demonstrated antigen retention after preparation methods.
Sigma-Aldrich
IgG from mouse serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Weigert′s iron hematoxylin kit, for nuclear staining in histology