Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.15944

Sigma-Aldrich

Methyl green zinc chloride double salt (C.I. 42590)

for microscopy Certistain®

Synonym(s):

Methyl green zinc chloride double salt (C.I. 42590), Ethyl green zinc chloride double salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H35BrClN3 · ZnCl2
CAS Number:
Molecular Weight:
653.24
MDL number:
UNSPSC Code:
41116130
EC Index Number:
230-415-4

Quality Level

form

solid

composition

Dye, ≥50% spectrophotometric assay

pH

4.6 (20 °C, 10 g/L in H2O)

mp

>300 °C

solubility

70 g/L

bulk density

300 kg/m3

εmax

≥810 at 630-635 nm in water at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C27H35N3.4ClH.Zn/c1-8-30(6,7)26-19-13-23(14-20-26)27(21-9-15-24(16-10-21)28(2)3)22-11-17-25(18-12-22)29(4)5;;;;;/h9-20H,8H2,1-7H3;4*1H;/q+2;;;;;+2/p-4

InChI key

ZKOGXCSOLMZRAZ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Methyl green zinc chloride double salt (C.I. 42590) for microscopy Certistain®, is a dry dye that is used for the preparation of a staining solution e.g. for DNA and RNA staining in methyl green-pyronine G staining method in histological sections of human origin.
As a Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (spectrophotometrically): ≥ 50 % C₂₇H₃₅BrClN₃
Identity
- Bromide:
passes test
- chloride: passes test
- Zinc: passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 630 - 635 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; water): ≥ 810
Crystal violet: ≤ 4 %
TLC-Test: passes test
Loss on drying (50 ° C/0.05 mm Hg/ 3h): ≤ 5 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service