Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.15942

Sigma-Aldrich

Malachite green oxalate (C.I. 42000)

for microscopy Certistain®

Synonym(s):

Malachite green oxalate (C.I. 42000)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H54N4O12
CAS Number:
Molecular Weight:
927.00
UNSPSC Code:
41116130
EC Index Number:
219-441-7

Quality Level

form

solid

composition

Dye, ≥90% spectrophotometric assay

pH

2.4 (24 °C, 10 g/L in H2O)

mp

159 °C

solubility

110 g/L

bulk density

400‑500 kg/m3

εmax

1730-1960 at 616-620 nm in water at 0.003 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/2C23H25N2.3C2H2O4/c2*1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;3*3-1(4)2(5)6/h2*5-17H,1-4H3;3*(H,3,4)(H,5,6)/q2*+1;;;/p-2

InChI key

CNYGFPPAGUCRIC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Malachite green oxalate (C.I. 42000) for microscopy Certistain®, is a dry dye that is used for the preparation of a staining solution that is used for differential spore staining in bacteriological samples of human origin.
As Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations.

Analysis Note

Dye content (spectrophotometrically): ≥ 90% (als C₄₆H₅₀N₄ *2 C₂HO₄ *C₂H₂O₄)
Identity
- oxalate:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 616 - 620 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; water): 1730 - 1960
TLC-Test: passes test
Loss on drying (50 °C / <1 hPa): ≤ 7%
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service