Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.15938

Sigma-Aldrich

Hematoxylin monohydrate (C.I.75290)

for microscopy Certistain®

Synonym(s):

Hematoxylin monohydrate (C.I.75290)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O6 · xH2O
Molecular Weight:
302.28 (anhydrous basis)
UNSPSC Code:
41116130
EC Index Number:
208-237-3
NACRES:
NA.21

Quality Level

form

solid

composition

Dye, ≥95% spectrophotometric assay

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

mp

140 °C (Elimination of water of crystallisation)

solubility

slightly soluble at 20 °C

bulk density

500 kg/m3

εmax

155-165 at 290-292 nm in acetonitrile at 0.05 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

General description

Hämatoxylin-Monohydrat (C.I. 75290) for microscopy Certistain®, is a dye for the specific staining of DNA in the nuclei. It is used in cytology and histology. Nuclei are stained dark blue, dark violet to black. Hematoxylins are mixed with trivalent positively charged metal salts and build up the so-called hematoxylin lakes, used for the selective staining of nuclei (DNA). Certistain® is the quality group for seleceted high quality dyes that fulfill all quality requirements for IVD and CE registration. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (spectrophotometrically): ≥ 95 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (acetonitrile): 290 - 292 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.05 g/l; acetonitrile): 155 - 165
Haematein (spectrophotometrically): ≤ 0.1 %
Water (according to Karl Fischer): 5 - 6 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jo Waaler et al.
Communications biology, 3(1), 196-196 (2020-04-26)
The development of immune checkpoint inhibitors represents a major breakthrough in cancer therapy. Nevertheless, a substantial number of patients fail to respond to checkpoint pathway blockade. Evidence for WNT/β-catenin signaling-mediated immune evasion is found in a subset of cancers including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service