Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.14770

Supelco

Manganese Test, photometric

photometric, 0.010-10.00 mg/L (Mn), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Manganese Test, photometric, 0.010-10.00 mg/L (Mn), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 250 tests (depending on package size)
sufficient for 500 tests

specific analyte(s)

manganese

measuring range

0.010-10.00 mg/L (Mn)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Formaldioxime)

storage temp.

15-25°C

General description

Method: photometric 0.010 - 10.00 mg/l Mn Spectroquant®
The method is analogous to DIN 38406-2.


This Spectroquant® Manganese Reagent Test allows the accurate quantification of the manganese(II) ions content in aqueous samples.
Method applied: In alkaline solution manganese(II) ions react with an oxime to form a red-brown complex which is determined photometrically.
The Spectroquant®Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Respiratory system, spleen

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service