Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.14663

Supelco

Oxygen Reaction Bottle

for use with MCOLORTEST®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NB.35

analyte chemical class(es)

oxygenates

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

We offer you useful accessories. The MQuant® oxygen reaction bottle (formally MColortest®) is used in combination with oxygen tests 1.11107 and 1.14662 to determine the oxygen concentration following the Winkler method. It is also included in the MQuant® compact laboratory for water testing 1.11151.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the importance of lab water quality on Chemical Oxygen Demand (COD) and Biochemical (or Biological) Oxygen Demand (BOD) testing in water and wastewater analysis. Find in-depth guidance to establish the optimal lab water solution for your environmental and process water testing.

Learn about the importance of lab water quality on Chemical Oxygen Demand (COD) and Biochemical (or Biological) Oxygen Demand (BOD) testing in water and wastewater analysis. Find in-depth guidance to establish the optimal lab water solution for your environmental and process water testing.

Learn about the importance of lab water quality on Chemical Oxygen Demand (COD) and Biochemical (or Biological) Oxygen Demand (BOD) testing in water and wastewater analysis. Find in-depth guidance to establish the optimal lab water solution for your environmental and process water testing.

Learn about the importance of lab water quality on Chemical Oxygen Demand (COD) and Biochemical (or Biological) Oxygen Demand (BOD) testing in water and wastewater analysis. Find in-depth guidance to establish the optimal lab water solution for your environmental and process water testing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service