Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.13900

Millipore

LiChroprep® RP-18 (40-63 µm)

for liquid chromatography

Synonym(s):

LiChroprep® RP-18

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41115711

material

spherical silica gel

Quality Level

particle size

40-63 μm

pore size

10 nm mean pore size

bulk density

410 kg/m3

application(s)

plasma purification

separation technique

reversed phase

storage temp.

no temp limit

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Carbon content: ≥ 15 %
Application test: passes test
Theoretical Plates (N/m) (Toluene): ≥ 1500
separation factor (Acetophenone/Cinnamic acid, selectivity test): 1.1 - 1.3
separation factor (Acetophenone/Phenol, selectivity test): 1.8 - 2.2
separation factor (1-Naphtol/Phenol, selectivity test): 3.9 - 4.4
Particle Size (d10): 31.0 - 37.0 µm
Particle Size (d50): 54.0 - 64.0 µm
Particle Size (d90): 95.0 - 110.0 µm
capacity factor (Toluene): 17.0 - 22.0

Legal Information

LICHROPREP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service