Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

1.13351

Supelco

Methanol

≥99.8% (GC), for preparative HPLC, for preparative liquid chromatography, for chromatography, Prepsolv

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-659-6
NACRES:
NB.21
grade:
for chromatography
for preparative HPLC
for preparative liquid chromatography
Assay:
≥99.8% (GC)
application(s):
food and beverages
pharmaceutical
bp:
64.7 °C (lit.)
vapor pressure:
128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Product Name

Methanol, for preparative chromatography Prepsolv

grade

for chromatography
for preparative HPLC
for preparative liquid chromatography

Quality Level

vapor density

1.11 (vs air)

vapor pressure

128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

product line

Prepsolv

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

725 °F

potency

5628 mg/kg LD50, oral (Rat)
17100 mg/kg LD50, skin (Rabbit)

expl. lim.

36 %

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.05% Water

evapn. residue

≤1.0 mg/L

transmittance

225 nm, ≥50%
265 nm, ≥98%

refractive index

n20/D 1.329 (lit.)

kinematic viscosity

0.54-0.59 cSt(20 °C)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Application


  • The composition of chemicals and anti-osteogenic properties in the volatile extracts from Homalomena gigantea rhizome: This study highlights methanol′s role in the extraction and analysis of volatile organic compounds from medicinal plants, providing insights valuable for researchers in pharmacology and natural product chemistry (Thanh et al., 2024).


  • A homochiral tartrate-bridged dinuclear chromium(III) complex anion with a resonance-assisted hydrogen bond for proton conduction: Methanol is used as a solvent in the synthesis of complex inorganic compounds, which are essential in developing new materials for energy applications, such as proton exchange membranes (Dunatov et al., 2024).



Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Evaporation Number: 1.9
Purity (GC): ≥ 99.8 %
Identity (IR): conforms
Evaporation residue: ≤ 1.0 mg/l
Water: ≤ 0.05 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Transmission (at 225 nm): ≥ 50 %
Transmission (from 265 nm): ≥ 98 %
Filtered by 0.2 µm filter

Legal Information

PREPSOLV is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service