Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.12416

Sigma-Aldrich

Tin(II) sulfate

LAB

Synonym(s):

Tin(II) sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O4SSn
CAS Number:
Molecular Weight:
214.77
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-302-2
NACRES:
NA.22

Quality Level

form

solid

potency

2207 mg/kg LD50, oral (Rat)

pH

1.6 (20 °C, 50 g/L in H2O)

bulk density

800 kg/m3

storage temp.

no temp limit

InChI

1S/H2O4S.Sn/c1-5(2,3)4;/h(H2,1,2,3,4);/q;+2/p-2

InChI key

OBBXFSIWZVFYJR-UHFFFAOYSA-L

Analysis Note

Assay (iodometric): ≥ 92.0 %
Identity (ICP-OES): passes test

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service