Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00822

Supelco

Fluoride Test, photometric

photometric, 0.02-2.00 mg/L (F), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Fluoride Test, photometric, 0.02-2.00 mg/L (F), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 250 tests

specific analyte(s)

fluoride

measuring range

0.02-2.00 mg/L (F)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (SPADNS method)

storage temp.

15-25°C

General description

Method: photometric, SPADNS 0.02 - 2.00 mg/l F Spectroquant®
This Spectroquant® Fluoride test allows the accurate quantification of fluoride ions in various waters. Fluoride ions occur on earth in several minerals, but are only present in trace quantities in bodies of water in nature. Fluoride ions react with part of a red zirconium-dye solution to form a colorless complex, which in turn results in a bleaching of the red color (SPADNS method). The concentration of the unconsumed zirconium-dye solution is subsequently determined photometrically.
The method is analogous to APHA 4500-F- D.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service