Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.11932

Millipore

Fermtech® Peptone from Soy

For Biotechnology, Suitable for Fermentation Technology

Synonym(s):

Fermtech® base materials for biotechnology, Peptone from Soy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

biological source

soybean

Quality Level

form

medium powder

loss

≤6.0% loss on drying, Tested at 105°C

pH

6.5-7.5 (25 °C)

application(s)

life science and biopharma
microbiology
pharmaceutical

storage temp.

15-25°C

suitability

nonselective for bacteria
nonselective for fungi

General description

Soy peptone is a non-animal peptone that contains a broad range of nutrients. It is characterized by its high concentration of vitamins and carbohydrates. Its nitrogen content combined with the naturally occurring vitamins and high carbohydrate content facilitates rapid and profuse growth of fastidious microorganisms, as well as resuscitation of sub-lethally injured microorganisms. Its high content of fermentable sugars makes soy peptone unsuitable for fermentation studies, such as in media used for the identification of microorganisms based on sugar fermentation.

Application

Soy peptone is used in cell culture, molecular genetic media, for microbiological assay procedures, and as a nutritive substrate for cultivation and resuscitation of a large variety of microorganisms.

Legal Information

FERMTECH is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service