Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.10046

Supelco

Total Hardness Test

greener alternative

colorimetric, water hardness 5-25 °d, for use with MQuant® StripScan App, water hardness 6-31 °e, water hardness 9-45 °f, MQuant®

Synonym(s):

Total Hardness chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

calcium, magnesium

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

water hardness measuring range

5-25 °d
6-31 °e
9-45 °f

compatibility

for use with MQuant® StripScan App

detection method

colorimetric

greener alternative category

storage temp.

15-25°C

General description

Method: Colorimetric, with test strips, >5 - >10 - >15 - >20 - >25°d, >6 - >13 - >19 - >25 - >31°e, >9 - >18 - >27 - >36 - >45°f, MQuant®

The MQuant® total hardness test strips can be used to measure water hardness. The hardness (total hardness) of a given water is due to its content of salts of the alkaline earth metals calcium, magnesium, strontium, and barium (“hardening constituents”, later compounds only in traces). In the presence of a green indicator the hardness ions Ca2+ und Mg2+ react with Titriplex® III to form a colorless, stable complex. The hardness ions that are present in excess relative to Titriplex® III react with the indicator to form a red complex. The reaction zones of the test strip, which are impregnated with differing amounts of Titriplex® III, thus assume a green to red color, depending on the hardness of the water sample. The hardness is measured semi-quantitatively by visual comparison of the reaction zones with the color rows of a color scale. For other sensitivities and graduations please consider the other MQuant® tests as well as our Reflectoquant® and Spectroquant® portfolio.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet
Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Linkage

Replaces: 10046-1; 10046

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service