Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.17924

Supelco

Chlorine Test Strips, colorimetric

greener alternative

0-500 mg/L (Cl₂), MQuant®

Synonym(s):

Chlorine chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Chlorine Test, colorimetric, 0-500 mg/L (Cl2), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

chlorine

reaction suitability

reaction type: Redox Reactions

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

0-500 mg/L (Cl2)

detection method

colorimetric

greener alternative category

storage temp.

2-8°C

General description

Method: Colorimetric with test strips, 0 - 25 - 50 - 100 - 200 - 500 mg/L Cl2, MQuant®

The MQuant® Chlorine test strips are suitable for a rapid determination of higher concentrations of chlorinated disinfectants, which are chiefly used in the food industry. On the test strip, free chlorine oxidizes an organic compound to a violet dye. The chlorine concentration is measured semiquantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. For a quantitative measurement or a measurement of total chlorine, please refer to our tests for chlorine from our Reflectoquant® or Spectroquant® portfolio.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Application


  • Analysis of free chlorine in aqueous solution at very low concentration with lateral flow tests.: This research discusses the use of lateral flow tests to measure free chlorine in water at very low concentrations, providing a sensitive method for chlorine detection in analytical chemistry (Schwenke et al., 2019).



Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for free Chlorine according to DIN EN ISO 7393

AQA Standard for free chlorine - Preparation of a standard solution for free chlorine

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service