Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09870

Supelco

Phosphate standard

1000 mg PO₄ (H₃PO₄ in H₂O) Titrisol®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

technique(s)

ion chromatography: suitable

pH

1.4 (20 °C in H2O)

density

1.02 g/cm3 at 20 °C

application(s)

environmental
food and beverages

format

single component solution

storage temp.

15-25°C

Analysis Note

content (PO₄³⁻): 1000 mg / Ampoule
Determination method: Alcalimetric titration.
(traceable to NIST - SRM 84k)
Accuracy of the method: +/- 2 mg

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service