Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09481

Supelco

ICP multi-element standard solution XIV

(11 elements in dilute hydrochloric acid) 100 mg/l: P, S, K; 20 mg/l: As, La, Li, Mo, Mn, Ni, Sc, Na Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

reference material

Quality Level

product line

Certipur®

technique(s)

ICP: suitable

pH

1 (20 °C in H2O)

bp

100 °C/1013 hPa

density

1.009 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

multi-component solution

storage temp.

15-25°C

Analysis Note

As (arsenic): 20 mg/l
K (potassium): 100 mg/l
La (Lanthanum): 20 mg/l
Li (Lithium): 20 mg/l
Mn (Manganese): 20 mg/l
Mo (Molydenium): 20 mg/l
Na (sodium): 20 mg/l
Ni (Nickel): 20 mg/l
P (Phosphorus): 100 mg/l
S (Sulfur): 100 mg/l
Sc (Scandium): 20 mg/l

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service