Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09242

Sigma-Aldrich

Kaiser′s glycerol gelatine

for microscopy

Synonym(s):

Glycerol gelatin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401

Quality Level

color

yellow

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Kaiser′s glycerol gelatin - for microscopy, is an aqueous mounting medium consisting of gelatine, glycerol, and phenol, and is used primarily in the mounting of stained histology specimens (e. g. HEMATOGNOST Fe - staining kit (Product number 1.12084). The pH of the product lies in the neutral range. Prior to use, the mounting medium is melted in the heating cabinet at approx. 60 °C and simply applied to the specimen using a glass rod. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

Aqueous mounting medium

Analysis Note

Suitability for microscopypasses test

Legal Information

HEMATOGNOST FE is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service