Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.08164

Supelco

Potassium chloride solution

UV-VIS Standard 4, according to Ph. Eur. stray light testing, Certipur®

Synonym(s):

UV-VIS Standard 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

Product Name

Potassium chloride solution, UV-VIS Standard 4, according to Ph. Eur. stray light testing, Certipur®

grade

analytical standard

Quality Level

Agency

according to Ph. Eur. stray light testing

product line

Certipur®

technique(s)

UV/Vis spectroscopy: suitable

application(s)

environmental
industrial qc

format

mixture

storage temp.

15-25°C

General description

for stray light testing according to Ph Eur Certipur®

Application

UV-VIS Standard 4: Potassium chloride solution as a reference material for checking the stray light of spectrophotometers in accordance with European Pharmacopoeia (Ph. Eur.) chapter 2.2.25 and American Society for Testing and Materials (ASTM) E 387.

Features and Benefits

  • It is a 12 g/L solution of potassium chloride in H2O
  • Conveniently packed in 10 mL glass ampules. Each box includes 3 single-use ampules
  • Stable solutions in glass ampoules
  • Accreditation according to ISO 17025
  • Traceability to standard reference material from the National Institute of Standards and Technology (NIST) and Physical-Technical Institute (PTB)
  • Includes a certificate of analysis (CoA)
  • Reliable and reproducible results

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service