Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.07204

Supelco

Certified sec. standard reference buffer solution

potassium tetraoxalate dihydrate, directly traceable to primary SRM from NIST/PTB pH(S) = 1.67₉ (25°C) Certipur®

Synonym(s):

Potassium tetraoxalate dihydrate buffer solution, pH 1.7 Certified Secondary Buffer Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.25

grade

certified reference material

Quality Level

Agency

complies with DIN 19266

product line

Certipur®

pH

1.7 (25 °C in H2O)

density

1.003 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The potassium tetraoxalate dihydrate certified secondary standard reference buffer solution is gravimetrically prepared using high-quality salt in high-purity water. Its pH value is measured by differential potentiometry with two platinum hydrogen electrodes against primary certified reference materials from NIST & PTB. The pH qualification is based on the 5-point calibration method, following DIN 19268, against certified primary reference standards as per DIN 19266.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

It can be used to calibrate, qualify, and monitor pH measuring instruments and as a control for testing other buffer solutions.

Features and Benefits

  • Available in a safe 100 mL high-density plastic bottle in solution format
  • Subjected to detailed stability studies as per ISO Guide 35
  • Product label carrying relevant information on order number, article description, lot number, expiry date and safety data, in accordance with ISO Guide 31
  • Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective quality management system
  • Measured and qualified in ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to SRMs from NIST and primary certified reference standards from PTB

Analysis Note

pH-value: 1.674 - 1.683
For complete data see the enclosed certificate

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service