Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.06310

Supelco

di-Sodium tetraborate

for analysis (ca. 85% Na₂B₄O₇)

Synonym(s):

di-Sodium tetraborate, Borax

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B4Na2O7 · nH2O (n=1.5-2)
CAS Number:
MDL number:
UNSPSC Code:
12352302
EC Index Number:
215-540-4
NACRES:
NA.21
grade:
for analytical purposes
form:
solid
solubility:
water: Ca. 25  g/L at 20 °C (68 °F)

grade

for analytical purposes

Quality Level

vapor pressure

20-°C ( 68 °F)

Assay

≥85.0% (acidimetric)

form

solid

pH

9-10 (20 °C, 25 g/L in H2O)

bp

1575 °C/1013 hPa (anhydrous)

mp

742 °C (anhydrous)

solubility

water: Ca. 25  g/L at 20 °C (68 °F)

density

2.35 g/cm3 at 26 °C

bulk density

680 kg/m3

anion traces

chloride (Cl-): ≤0.002%
phosphate (PO43-): ≤0.002%
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.01%
Fe: ≤0.001%
heavy metals (as Pb): ≤0.002%

storage temp.

2-30°C

Application


  • Micellar electrokinetic chromatography for separation of a mixture of coptis alkaloids, scute flavonoids, and rhubarb anthraquinones and bianthrones.: This study by Chang and Sun investigates the use of micellar electrokinetic chromatography (MEKC) to separate complex mixtures of bioactive compounds. The research highlights the effectiveness of di-Sodium tetraborate in optimizing the separation process, showcasing its utility in analytical chemistry for the precise analysis of natural products (Chang et al., 2006).

Analysis Note

Assay (acidimetric): ≥ 85.0 %
Chloride (Cl): ≤ 0.002 %
Phosphate (PO₄): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.002 %
Ca (Calcium): ≤ 0.01 %
Fe (Iron): ≤ 0.001 %

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service