Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 g | Check Cart for Availability | $236.00 $177.00 |
About This Item
grade
certified reference material
Quality Segment
agency
complies with DIN 19266
vapor pressure
0.213 hPa ( 20 °C)
product line
Certipur®
potency
2660 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rabbit)
pH
9.2 (20 °C, 47 g/L in H2O)
mp
75 °C (Elimination of water of crystallisation)
solubility
49.74 g/L
density
1.72 g/cm3 at 20 °C
bulk density
750 kg/m3
application(s)
environmental
food and beverages
general analytical
industrial qc
pharmaceutical
format
neat
storage temp.
15-25°C
SMILES string
[Na+].[Na+].B([O-])(OB([O-])OB=O)OB=O.O.O.O.O.O.O.O.O.O.O
InChI
1S/B4O7.2Na.10H2O/c5-1-9-3(7)11-4(8)10-2-6;;;;;;;;;;;;/h;;;10*1H2/q-2;2*+1;;;;;;;;;;
InChI key
OJPFLODMGUZMKD-UHFFFAOYSA-N
General description
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Application
Features and Benefits
- The product is delivered with a comprehensive, batch-specific Certificate of Analysis showing the relevant data for the quality management system.
The pH value of this Certipur® buffer solution is measured using differential potentiometry with two platinum hydrogen electrodes against NIST & PTB primary SRMs [pH(S)=9.180 (25 °C)].
- It is a Certified Reference Material (CRM) offered in neat format.
- Intended for extremely precise calibration, qualification and monitoring of pH instruments and for testing other buffer solutions.
Analysis Note
Identity: passes test
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| format neat | format - | format - | format - |
| mp 75 °C (Elimination of water of crystallisation) | mp 75 °C (Elimination of water of crystallisation) | mp 75 °C (Elimination of water of crystallisation) | mp - |
| product line Certipur® | product line EMPROVE® ESSENTIAL | product line - | product line - |
| grade certified reference material | grade - | grade ACS reagent | grade - |
| application(s) environmental | application(s) liquid formulation | application(s) - | application(s) - |
| pH 9.2 (20 °C, 47 g/L in H2O) | pH 9.2 (20 °C, 47 g/L in H2O) | pH 9.2 (20 °C, 47 g/L in H2O) | pH 9.0-9.6 (20 °C, 5%) |
Still not finding the right product?
Explore all of our products under di-Sodium tetraborate decahydrate
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Repr. 1B
Storage Class
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





