Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.06260

Supelco

Sodium

rod diameter 2.5 cm (protective liquid: paraffin oil)

Synonym(s):

Sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na
CAS Number:
Molecular Weight:
22.99
MDL number:
UNSPSC Code:
12141806
EC Index Number:
231-132-9
NACRES:
NA.21

vapor pressure

1.6 hPa ( 400 °C)

Quality Level

form

solid

autoignition temp.

>115 °C

impurities

≤0.01% Chloride (Cl)

bp

889 °C/1013 hPa

mp

97.8 °C

density

0.97 g/cm3 at 20 °C

cation traces

Ca: ≤0.1%
K: ≤0.05%

storage temp.

2-30°C

InChI

1S/Na/q+1

InChI key

FKNQFGJONOIPTF-UHFFFAOYSA-N

Analysis Note

Identity: passes test
Chloride (Cl): ≤ 0.01 %
Ca (Calcium): ≤ 0.1 %
K (Potassium): ≤ 0.05 %

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

179.6 °F

Flash Point(C)

82 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service