Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71172

Sigma-Aldrich

Sodium

in kerosene, pieces (large), ≥99.8% (sodium basis)

Synonym(s):

Sodium metal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na
CAS Number:
Molecular Weight:
22.99
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 440 °C)

Assay

≥99.8% (sodium basis)

form

pieces (large)

quality

in kerosene

reaction suitability

reagent type: reductant

resistivity

4.69 μΩ-cm, 20°C

bp

883 °C (lit.)

mp

97.8 °C (lit.)

anion traces

chloride, bromide (as Cl)-: ≤20 mg/kg

cation traces

Ca: ≤400 mg/kg
K: ≤300 mg/kg

SMILES string

[Na]

InChI

1S/Na

InChI key

KEAYESYHFKHZAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium metal (in kerosene) has been used in the electrochemical reformation of levulinic acid to form 5-acetyl-2,9-decanedione. It may be used as a drying agent for the purification of tetrahydrofuran (THF).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drying of organic solvents: quantitative evaluation of the efficiency of several desiccants.
Williams D
The Journal of Organic Chemistry, 75(24), 8351-8354 (2010)
Electrochemical synthesis of a novel compound, 5-acetyl-2, 9-decanedione, and theoretical analysis of its lithium ion complex.
Cabasso I
Royal Society of Chemistry Advances, 2(26), 9998-10006 (2012)
Highly stable multi-anchored magnetic nanoparticles for optical imaging within biofilms
Stone R C
Journal of Colloid and Interface Science, 459, 175-182 (2015)
Andrew Mente et al.
The New England journal of medicine, 371(7), 601-611 (2014-08-15)
Higher levels of sodium intake are reported to be associated with higher blood pressure. Whether this relationship varies according to levels of sodium or potassium intake and in different populations is unknown. We studied 102,216 adults from 18 countries. Estimates
Supriya S Jalimarada et al.
Investigative ophthalmology & visual science, 54(6), 4330-4340 (2013-06-08)
Mutations in SLC4A11, a member of the SLC4 superfamily of bicarbonate transporters, give rise to corneal endothelial cell dystrophies. SLC4A11 is a putative Na⁺ borate and Na⁺:OH⁻ transporter. Therefore we ask whether SLC4A11 in corneal endothelium transports borate (B[OH]₄⁻), bicarbonate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service