Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.06202

Supelco

1-Naphtholbenzein

pH indicator, Reag. Ph Eur

Synonym(s):

1-Naphtholbenzein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H18O2
CAS Number:
Molecular Weight:
374.43
UNSPSC Code:
12352115
EC Index Number:
205-656-3
NACRES:
NB.21

product name

1-Naphtholbenzein, indicator Reag. Ph Eur

Agency

reag. Ph. Eur.

Quality Level

form

solid

loss

≤7% loss on drying, 105°C

visual transition interval

8.0-9.6, apricot to turquoise

mp

243-245 °C

bulk density

120‑300 kg/m3

λmax

622-626 nm (perchloric acid (0,1 mol/l) in acetic acid)

storage temp.

2-30°C

InChI

1S/C27H18O2/c28-25-16-14-23(19-10-4-6-12-21(19)25)27(18-8-2-1-3-9-18)24-15-17-26(29)22-13-7-5-11-20(22)24/h1-17,28H/b27-24+

InChI key

VDDWRTZCUJCDJM-SOYKGTTHSA-N

Application


  • Use of turbidimeter for measurement of solid catalyst system component in a reactor feed.: This study discusses the application of a turbidimeter to measure the concentration of solid catalyst components in a reactor feed, highlighting its significance in optimizing catalytic processes (Elsevier, 2020).



Analysis Note

Identity (IR-spectrum): passes test
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax.(perchloric acid (0,1 mol/l) in acetic acid): 622 - 626 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l;perchloric acid (0,1 mol/l) in acetic acid;calculated on dried substance): 640 - 950
Transition range: pH 8,0 - pH 9,6; apricot - turquoiseblue
Loss on Drying (105°C): ≤ 7 %
Suitability as indicator (for non-aqueous titration): passes test

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service