Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.05411

Millipore

GranuCult® prime Reinforced clostridial medium (RCM)

according to EP, according to USP, according to JP, for Clostridium spp., suitable for microbiology

Synonym(s):

Clostridium differential medium, RCA Agar, RCM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

according to EP
according to JP
according to USP

Quality Level

sterility

non-sterile

form

granular

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® prime

storage condition

dry media (Keep tightly closed)

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Clostridium spp.

General description

GranuCult® prime Reinforced clostridial medium (RCM) be used to initiate growth from small inoculum and to obtain the highest viable count of Clostridia. It can be used as the conventional medium for studies of spore-forming anaerobes, especially Clostridium butyricum in cheese, and for the enumeration and isolation of Clostridia. Other spore-forming anaerobes like streptococci and lactobacilli grow as well on this media. Casein hydrolysate, beef extract, yeast extract, and peptone provide nitrogen, vitamins, amino acids, and carbon for growth. Dextrose is a fermentable sugar, and sodium chloride ensures osmotic balance. The medium is free from inhibitors and contains cysteine as a reducing agent. Sodium acetate is the buffering agent. Polymyxin B can be added to inhibit Gram-negative bacteria and to make the medium more selective. Culture conditions include incubation at 35-37°C for 40-48 hours in an anaerobic atmosphere.

Application

GranuCult® prime Reinforced clostridial medium (RCM) is suitable for the cultivation and enumeration of clostridia, other anaerobes, and facultative microorganisms in foodstuffs and other materials.

Features and Benefits

  • GranuCult® offers superior granulated culture media
  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment
  • Excellent wettability, solubility, and free-flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allows many supplements to be included, with no need to add these separately

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish
pH-value (25 °C): 6.6 - 7.0
Typical composition (g/litre): Meat extract 10.0; Peptones 10.0; Yeast extract 3.0; D(+)Glucose 5.0; Starch 1.0; Sodium chloride 5.0; Sodium acetate 3.0; L-Cysteinium chloride 0.5; Agar-agar 0.5
Growth promotion test in accordance with the harmonised method of EP, USP and JP.
Inoculum on reference medium (Clostridium sporogenes ATCC 11437): ≤ 100
Inoculum on reference medium (Clostridium sporogenes ATCC 19404 (WDCM 00008)): ≤ 100
Inoculum on reference medium (Clostridium perfringens ATCC 10543): ≤ 100
Inoculum on reference medium (Clostridium novyi ATCC 17861): ≤ 100
Inoculum on reference medium (Escherichia coli ATCC 25922 (WDCM 00013)): ≤ 100
Growth (Clostridium sporogenes ATCC 11437): +
Growth (Clostridium sporogenes ATCC 19404 (WDCM 00008)): +
Growth (Clostridium perfringens ATCC 10543): +
Growth (Clostridium novyi ATCC 17861): +
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): +
Incubation: 48 hrs.; 30-35°C; Clostridia: anaerobic
The Information above is current at this time of publication and is subject to change without notice (except for customers holding a change control agreement with our company). The information/format can be altered at any time and inany way if internal company matters or Standard Regulationsdo say so.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service