Skip to Content
MilliporeSigma

1.05281

Lysozyme

(from egg white) ≥ 30 000 FIP-U/mg cryst. (hydrochloride) for biochemistry EC 3.2.1.17

Synonym(s):

Lysozyme, Muramidase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
EC Index Number:
232-620-4
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

egg white

Quality Segment

form

solid

pH

3.5 (20 °C in H2O, Aqueous solution)

solubility

300 g/L

storage temp.

2-8°C

SMILES string

N1([C@@H](CCC1)C(=O)N[C@@H](CCC(=O)N)C(=O)NCC(=O)N[C@@H]([C@H](CC)C)C(=O)N[C@@H](CCCN\C(=N\[H])\N)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc4c5c([nH]c4)cccc5)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc2c3c([nH]c2)cccc3)C(=O)N[C@@H](CCCN\C(=N\[H])\N)C(=O)N[C@@H]

InChI

1S/C99H159N37O23/c1-11-50(8)77(132-72(139)46-120-81(145)63(32-33-70(101)137)124-88(152)69-31-21-39-136(69)93(157)68(43-73(140)141)131-84(148)62(29-19-37-116-98(109)110)125-90(154)75(48(4)5)135-91(155)76(49(6)7)133-80(144)57(100)24-16-34-113-95(103)104)92(156)126-60(27-17-35-114-96(105)106)82(146)121-51(9)78(142)129-66(41-54-45-119-59-26-15-13-23-56(54)59)87(151)134-74(47(2)3)89(153)122-52(10)79(143)128-65(40-53-44-118-58-25-14-12-22-55(53)58)85(149)123-61(28-18-36-115-97(107)108)83(147)130-67(42-71(102)138)86(150)127-64(94(158)159)30-20-38-117-99(111)112/h12-15,22-23,25-26,44-45,47-52,57,60-69,74-77,118-119H,11,16-21,24,27-43,46,100H2,1-10H3,(H2,101,137)(H2,102,138)(H,120,145)(H,121,146)(H,122,153)(H,123,149)(H,124,152)(H,125,154)(H,126,156)(H,127,150)(H,128,143)(H,129,142)(H,130,147)(H,131,148)(H,132,139)(H,133,144)(H,134,151)(H,135,155)(H,140,141)(H,158,159)(H4,103,104,113)(H4,105,106,114)(H4,107,108,115)(H4,109,110,116)(H4,111,112,117)/t50-,51-,52-,57-,60-,61-,62-,63-,64-,65-,66-,67-,68-,69-,74-,75-,76-,77-/m0/s1

InChI key

ZJCXKXFAOCLSRV-RYLVIXIQSA-N

General description

(from egg white) ≥ 30 000 FIP-U/mg cryst. (hydrochloride) for biochemistry EC 3.2.1.17

Application

Lysozyme (from egg white) ≥ 30 000 FIP-U/mg cryst. (hydrochloride) for biochemistry EC 3.2.1.17. CAS 9001-63-2, pH 3.5 (H₂O, 20 °C) Aqueous solution.

Analysis Note

Activity (Micrococcus luteus (FIP-Substrate); Lysozym (FIP-Standard); pH 7,0; 25,0 °C)≥ 30000 FIP-U/mgThe information provided does not imply the suitability of the product for any particular application. It is customer’s sole responsibility, prior to use, to determine that the product is suitable and permitted for the customer’s intended use and application.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library