Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.05238

Supelco

Ion exchanger Dowex 50 WX 4

(strongly acidic cation exchanger), H⁺ form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41115711

Quality Level

form

beads

autoignition temp.

>500 °C

technique(s)

LPLC: suitable

matrix

PS-DVB

matrix active group

SCX

particle size

180-500 μm

capacity

640 - 720  g/kg water absorbing capacity

density

1.040 g/cm3 at 20 °C

separation technique

cation exchange

storage temp.

15-25°C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Analysis Note

Description: moist beige spherules
Total capacity (wet): ≥ 1.1 mol/l
Particle size (0.18-0.5 mm): ≥ 70.0 %

Legal Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service