Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.05092

Supelco

Potassium peroxodisulfate

for analysis (<= 0.001% N) EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Potassium peroxodisulfate, Potassium persulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K2O8S2
CAS Number:
Molecular Weight:
270.32
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-781-8
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (iodometric)

form

solid

potency

802 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

pH

3.2 (20 °C, 50 g/L in H2O)

mp

100 °C (decomposition)

density

2.48 g/cm3 at 20 °C

bulk density

1150 kg/m3

anion traces

chloride (Cl-): ≤0.001%

cation traces

Fe: ≤0.0005%
Mn: ≤0.0001%
N: ≤0.001%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

InChI

1S/2K.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2

InChI key

USHAGKDGDHPEEY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (iodometric): ≥ 99.0 %
Insoluble matter: ≤ 0.005 %
Entire chloride: ≤ 0.001 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
Mn (Manganese): ≤ 0.0001 %
Corresponds to ACS, Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service