Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.04350

Millipore

L-Histidine monohydrochloride monohydrate

for biochemistry

Synonym(s):

His, L-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2 · HCl · H2O
CAS Number:
Molecular Weight:
209.63
Beilstein:
4168261
MDL number:
UNSPSC Code:
12352209
EC Index Number:
211-438-9
NACRES:
NA.21

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

crystalline powder

pH

3.5-4.5 (20 °C, 100 g/L in H2O)

mp

254 °C (dec.) (lit.)

solubility

149.55 g/L

density

1.49 g/cm3 at 20 °C

bulk density

770 kg/m3

storage temp.

2-30°C

SMILES string

O.Cl.N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m0../s1

InChI key

CMXXUDSWGMGYLZ-XRIGFGBMSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (alkalimetric, calculated on dried substance): ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white crystalline powder or colorless crystals
Appearance of solution (50 g/l CO₂-free water): clear and colorless
Spec. rotation (α 20/D, 110 g/l, hydrochloric acid 120 g/l, calc. on dried substance): +9.2 to +10.6
Sulfate (SO₄): ≤ 0.02 %
Heavy metals (as Pb): ≤ 0.001 %
As (Arsenic): ≤ 0.0005 %
Ca (Calcium): ≤ 0.001 %
Co (Cobalt): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
NH₄ (Ammonium): ≤ 0.01 %
Zn (Zinc): ≤ 0.0005 %
Foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances: ≤ 0.1 %
Loss on drying (150 °C): 7.0 - 10.0 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service