Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.04191

Supelco

Glyoxalbis(2-hydroxyanil)

GR for analysis Reag. Ph Eur

Synonym(s):

Glyoxalbis(2-hydroxyanil), 1,2-Bis(2-hydroxyphenylimino)ethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N2O2
CAS Number:
Molecular Weight:
240.26
UNSPSC Code:
12352116
EC Index Number:
214-560-0
NACRES:
SE.11

Agency

reag. Ph. Eur.

Quality Level

form

solid

mp

200 °C

bulk density

220 kg/m3

storage temp.

15-25°C

InChI

1S/C14H12N2O2/c17-13-7-3-1-5-11(13)15-9-10-16-12-6-2-4-8-14(12)18/h1-10,15,17H/b10-9+,16-12?

InChI key

OMJNMRJMNUREOJ-KOBPNRPCSA-N

Application


  • A new coordination mode of the photometric reagent glyoxalbis(2-hydroxyanil).: This study introduces a novel coordination mode for glyoxalbis(2-hydroxyanil), describing its use as a bis-bidentate bridging ligand in various ruthenium complexes. The research covers redox-active complexes and includes a radical-bridged diruthenium(III) and a unique Ru(III)/Ru(IV) intermediate, demonstrating potential applications in photometric analysis and redox chemistry (Kar et al., 2005).


Analysis Note

Assay (ex N): ≥ 97
Identity (IR-spectrum): passes test
Melting point: about 200
Sulfated ash: ≤ 0.1 %
Loss on Drying (105°C): ≤ 0.5 %
Suitability for metal determination: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service