Skip to Content
MilliporeSigma

1.01192

di-Ammonium oxalate monohydrate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

di-Ammonium oxalate monohydrate, Oxalic acid ammonium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(NH4)2C2O4 · H2O
CAS Number:
Molecular Weight:
124.10 (anhydrous basis)
UNSPSC Code:
12352107
EC Index Number:
214-202-3
NACRES:
NA.21
MDL number:
Assay:
99.5-101.0% (manganometric)
Grade:
ACS reagent
Form:
solid
Solubility:
41.8 g/L (anhydrous substance)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

agency

reag. ISO, reag. Ph. Eur.

vapor pressure

<1 hPa ( 20 °C)

product line

EMSURE®

assay

99.5-101.0% (manganometric)

form

solid

impurities

≤0.005% Insoluble matter

ign. residue

≤0.02% (as sulfate)

pH

6.5 (20 °C, 41.8 g/L in H2O)

mp

>123 °C (decomposition)

solubility

41.8 g/L (anhydrous substance)

density

1.5 g/cm3 at 20 °C

bulk density

480 kg/m3

anion traces

chloride (Cl-): ≤0.0005%, nitrate (NO3-): ≤0.002%, sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.001%, Fe: ≤0.0002%, K: ≤0.001%, Mg: ≤0.001%, Na: ≤0.001%, heavy metals: ≤0.0005% ((ACS))

storage temp.

2-30°C

SMILES string

N.N.OC(=O)C(=O)O.O

InChI

1S/C2H2O4.2H3N.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);2*1H3;1H2

InChI key

MSMNVXKYCPHLLN-UHFFFAOYSA-N

Analysis Note

Assay (manganometric): 99.5 - 101.0 %


Insoluble matter: ≤ 0.005 %


Chloride (Cl): ≤ 0.0005 %


Nitrate (NO₃): ≤ 0.002 %


Sulfate (SO₄): ≤ 0.002 %


heavy metals (ACS) ≤ 0.0005%


Ca (Calcium): ≤ 0.001 %


Fe (Iron): ≤ 0.0002 %


K (Potassium): ≤ 0.001 %


Mg (Magnesium): ≤ 0.001 %


Na (Sodium): ≤ 0.001 %


Residue on ignition (as sulfate): ≤ 0.02 %


Corresponds to ACS,ISO,Reag. Ph Eur

Other Notes

Replaces: AX1325; AX1325-2; AX1325-20; AX1325-1

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under di-Ammonium oxalate monohydrate


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

hcodes

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library