Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.04172

Sigma-Aldrich

Toluidine blue O (C.I.52040)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16ClN3S
CAS Number:
Molecular Weight:
305.83
MDL number:
UNSPSC Code:
41116130
NACRES:
NA.21

form

solid ()

Quality Level

composition

Dye, ≥85% spectrophotometric assay

pH

2.8 (25 °C in H2O)

solubility

30 g/L

bulk density

410 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C15H16N3S.ClH/c1-9-6-13-15(8-11(9)16)19-14-7-10(18(2)3)4-5-12(14)17-13;/h4-8H,16H2,1-3H3;1H/q+1;/p-1

InChI key

HNONEKILPDHFOL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Toluidine blue O (C.I. 52040) for microscopy Certistain® is adry dye that is used for the preparation of a staining solution for staining nuclei and for visualization of metachromasia. As a CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications.

Application

for microscopy Certistain®

Analysis Note

Dye content (Diaminophenothiazins, spectrophotometrically): ≥ 85 %
Identity
- chloride:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 628 - 633 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; water): 1330 - 1570
TLC-Test: passes test
Loss on drying (110 °C): ≤ 8 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service