Skip to Content
MilliporeSigma
All Photos(3)

Documents

1.04143

Millipore

Singlepath® Campylobacter Kit

suitable for immunological and lateral flow essay testing in Food and Beverages applications

Synonym(s):

Rapid Campylobacter Food Test

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NA.24

product name

Singlepath® Campylobacter, suitable for lateral flow assay, for Campylobacter spp., input: food(s)

Agency

according to AOAC® PTM

Quality Level

product line

Singlepath®

shelf life

15 mo.

packaging

pack of 25 strips

technique(s)

lateral flow assay: suitable
microbe id | immunological: suitable

input

sample type food(s)

application(s)

food and beverages
pathogen testing

storage temp.

2-8°C

suitability

Campylobacter spp.

General description

Singlepath® Campylobacter is a rapid diagnostic test, designed to detect Campylobacter bacteria in various food samples. Campylobacter is a genus of bacteria that can cause human infections, leading to symptoms such as diarrhea, fever, and abdominal cramps.The Singlepath® Campylobacter is an immunochromatographic rapid test based on gold-labeled antibodies to detect the presence of a specific antigen, in this case, the Campylobacter bacteria. The test includes a nitrocellulose membrane strip that is coated with antibodies specific to Campylobacter antigens. When the sample is applied to the test strip, any Campylobacter antigens present will bind to the antibodies on the strip, developing a visible test line. The test takes approximately 20 minutes to complete, and the results are interpreted based on the presence or absence of the test line. No additional equipment expenditure is needed.

Application

Singlepath® Campylobacter test is used to detect Campylobacter contaminations and growth in food samples. This product can also be used for pathogen testing in industrial microbiology. The test has been validated and approved by AOAC-PTM.

Features and Benefits

  • Rapid and reliable method
  • High sensitivity and specificity compared to conventional culture methods
  • Can be performed without the need for specialized equipment or personnel.

Linkage

Replaces: 1.04143

Analysis Note

Performance test (Campylobacter coli ATCC 33559 (WDCM 00072)): passes test
Performance test (Campylobacter jejuni ATCC 33560): passes test
Performance test (Escherichia coli NCTC 9001): passes test

Legal Information

AOAC is a registered trademark of AOAC International
SINGLEPATH is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Related Content

Detect pathogens rapidly using our lateral flow immunological tests which act as mini-laboratories for identifying the presence of specific pathogens.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service