Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.00079

Millipore

Bolton Broth Selective Supplement

according to ISO 10272-1, for Campylobacter spp., pkg of 10 vials, For the preparation of Bolton Selective Enrichment Broth

Synonym(s):

Campylobacter Selective Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

Agency

according to ISO 10272-1

Quality Level

reg. compliance

FDA -BAM

form

lyophilized powder

composition

Amphotericin B, 5 mg
Cefoperazone, 10 mg
Trimethoprim, 10 mg
Vancomycin, 10 mg

packaging

pkg of 10 vials

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology
pathogen testing

storage temp.

2-8°C

suitability

Campylobacter spp.
Listeria spp.

General description

Bolton Broth Selective Supplement is a mixture of four different antibiotics in lyophilized form. Vancomycin, Cefoperazone, and Trimethoprim inhibit the growth of Gram-positive and Gram-negative bacteria. Amphotericin B largely reduces the growth of Yeasts and Molds.

Application

Bolton Broth Selective Supplement is an additive used during the preparation of Bolton Selective Enrichment Broth for the enrichment of Campylobacter from foods.

Linkage

Replaces: 1.00079.0017

Analysis Note

Growth promotion test according to ISO 11133
Mixed inoculum of Campylobacter, E.coli and P.mirabilis.
Additionaly monoculture of E.coli and P.mirabilis.
Afterwards confirmation on mCCDA resp. Tryptic soy agar
Inoculum on reference medium
- Campylobacter coli ATCC 43478 (WDCM 00004):
10 - 100
- Campylobacter jejuni ATCC 33291 (WDCM 00005): 10 - 100
- Campylobacter jejuni ATCC 29428 (WDCM 00156): 10 - 100
- Escherichia coli ATCC 25922 (WDCM 00013): ≥ 1000
- Escherichia coli ATCC 8739 (WDCM 00012): ≥ 1000
- Proteus mirabilis ATCC 29906 (WDCM 00023): ≥ 1000
Growth on mCCDA
- Campylobacter coli ATCC 43478 (WDCM 00004):
>10 CFU
- Campylobacter jejuni ATCC 33291 (WDCM 00005): >10 CFU
- Campylobacter jejuni ATCC 29428 (WDCM 00156): >10 CFU
Growth on Tryptic soy agar
- Escherichia coli ATCC 25922 (WDCM 00013):
no growth
- Escherichia coli ATCC 8739 (WDCM 00012): no growth
- Proteus mirabilis ATCC 29906 (WDCM 00023): no growth
Colony colour
- Campylobacter coli ATCC 43478 (WDCM 00004):
greyish
- Campylobacter jejuni ATCC 33291 (WDCM 00005): greyish
- Campylobacter jejuni ATCC 29428 (WDCM 00156): greyish
Incubation:
Bolton broth: 5 hours at 44°C, afterwards 44 hours at 41.5°C, Anaerocult C
Confirmation on mCCDA: at least 44 hours at 41.5°C, Anaerocult C
Confirmation on Tryptic soy agar: 24 hours at 35°C, aerobic

Pictograms

Health hazard

Signal Word

Danger

Hazard Classifications

Repr. 2 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service