Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.03870

Millipore

GranuCult® prime Brain Heart Infusion Agar

acc. FDA-BAM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Quality Level

form

powder

technique(s)

microbe id | specific enzyme detection: suitable
microbiological culture: suitable

application(s)

food and beverages
pharmaceutical

suitability

Enterococcus spp.
Haemophilus spp.
Pneumococcus spp.
Staphylococcus spp.
Streptococcus spp.

General description

GranuCult® prime Brain Heart Infusion (BHI) Agar is a nutrient rich complex medium that is designed to cultivate and maintain fastidious microorganisms from food and animal feed, water and other samples.This culture medium can be used as a general-purpose medium, it contains no inhibitors or dyes and a broad spectrum of bacteria and fungi are able to grow on it. The highly nutritious substrate prepared from extracts of brain and heart, and peptones provide nitrogen, vitamins, amino acids and carbon sources. Glucose is an additional carbon source whilst disodium phosphate acts as a buffer. Sodium chloride maintains the osmotic balance and agar is the solidifying agent.

Application

GranuCult® prime Brain Heart Infusion (BHI) Agar is a nutrient rich complex medium.
Brain Heart Infusion (BHI) agar poured in slants is used for maintenance of pure cultures of microorganisms.

Brain Heart Infusion (BHI) agar is less suited for identifying hemolytic forms when blood has been added due to its glucose content.

According to APHA Compendium of Methods for microbiological examination of foods, BHI agar may be supplemented with vancomycin for the selective screening of vancomycin-resistant enterococci (VRE) from food samples

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service