Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.02769

Sigma-Aldrich

Copper(II) oxide

Technipur®

Synonym(s):

Copper monoxide, Copper oxide, Cupric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.21

Quality Level

pH

7 (20 °C, 50 g/L in H2O, slurry)

mp

1326 °C (decomposition)

SMILES string

[Cu]=O

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Copper(II) oxide (CuO) is a transition metal oxide and a p-type semiconductor commonly used as thermal conducting material, catalyst, gas sensor, magnetic storage media and varistors. It has a narrow bandgap of 1.2 eV and is used for optoelectronics, semiconductor and solar cell applications.

Application

Copper(II) oxide can be used as:
  • A dielectric material applicable in the microelectronic industry.
  • A hydrogen sulfide gas sensing material in biogas plants and natural gas facilities.
  • An induced-multiferroic material having a high TC temperature applicable in the manufacturing of superconductors.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Specific, trace gas induced phase transition in copper (II) oxide for highly selective gas sensing
Kneer J, et al.
Applied Physics Letters, 105(7), 073509-073509 (2014)
Cupric oxide as an induced-multiferroic with high-TC
Kimura T, et al.
Nature Materials, 7(4), 291-294 (2008)
Copper oxide nanomaterials prepared by solution methods, some properties, and potential applications: a brief review
Tran TH and Nguyen VT
International Scholarly Research Network (2014)
CuO and Co3O4 nanoparticles: synthesis, characterizations, and Raman spectroscopy
Rashad M, et al.
Journal of Nanomaterials, 2013 (2013)
Copper (II) oxide as a giant dielectric material
Sarkar S, et al.
Applied Physics Letters, 89(21), 212905-212905 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service