Skip to Content
MilliporeSigma

1.02767

Copper oxide

wire about 0.65 x 6 mm for elementary analysis (surface: CuO, core: Cu2O) ACS

Synonym(s):

Copper oxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
UNSPSC Code:
12352303
EC Index Number:
215-269-1
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

assay

(oxygen demand as CuO)- ≥ 55.0 %

form

solid

weight

=79.54 g/mol

mp

1300 °C

density

6.2 g/cm3 at 20 °C

bulk density

2200 kg/m3

cation traces

C: ≤0.002%, N: ≤0.002%

storage temp.

2-30°C

SMILES string

O=[Cu]

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Application


  • Systematic Genotoxicity Assessment of Metal Oxide Nanoparticles: Investigates the DNA damaging and clastogenic potential of various metal oxide nanoparticles including copper oxide, highlighting concerns and applications in toxicology (Solorio-Rodriguez SA et al., 2024).

  • Gold-copper Oxide Core-shell Nanoparticles: Investigates the effect of pH on shell stability and provides mechanistic insights into shell formation, critical for semiconductor and catalytic applications (Bartolucci SF et al., 2024).

Analysis Note

Assay (oxygen demand as CuO): ≥ 55.0 %
Chloride (Cl): ≤ 0.005 %
Total carbon (C): ≤ 0.002 %
Total nitrogen (N): ≤ 0.002 %
Total sulfur (as SO₄): ≤ 0.005 %


Still not finding the right product?

Explore all of our products under Copper oxide


pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library