Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.02531

Supelco

Cyanide Cell Test (free)

photometric, 0.010-0.500 mg/L (CN-), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116

product line

Spectroquant®

Quality Level

specific analyte(s)

cyanide

measuring range

0.010-0.500 mg/L (CN-)

technique(s)

photometry: suitable

detection method

photometric

General description

Method: photometric 0.010 - 0.500 mg/l CN⁻ Spectroquant®
he method corresponds to DIN 38405-13 and is analogous to ISO 6703, EPA 335.2, APHA 4500-CN-E, and ASTM D2036-09 D.

This Spectroquant Cyanide Cell Test allows the accurate quantification of the free cyanide content in aqueous samples.

Method applied: Cyanide ions react with a chlorinating agent to form cyanogen chloride which together with 1,3-dimethylbarbituric acid forms a violet dye (pyridine-free König reaction) that is determined photometrically.

The Spectroquant Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.

All Spectroquant Cell and Reagent Tests are equipped with the unique Live ID which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service