Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.02521

Supelco

Sodium hexanitrocobaltate(III)

[sodium cobalt(III)nitrite] for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Sodium hexanitrocobaltate(III), Cobalt(III) sodium nitrite, Sodium cobaltinitrite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3[Co(NO2)6]
CAS Number:
Molecular Weight:
403.94
MDL number:
UNSPSC Code:
12352302
EC Index Number:
237-077-7
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

form

powder

impurities

≤0.02% Insoluble matter

pH

5 (20 °C, 100 g/L in H2O)

mp

220 °C

bulk density

480 kg/m3

anion traces

chloride (Cl-): ≤0.005%

cation traces

Fe: ≤0.002%

storage temp.

2-30°C

InChI

1S/Co.6HNO2.3Na/c;6*2-1-3;;;/h;6*(H,2,3);;;/q;;;;;;;3*+1/p-6

InChI key

MUTOTFDDGIZNIK-UHFFFAOYSA-H

Related Categories

Application


  • One-pot synthesis of graphene-cobalt hydroxide composite nanosheets (Co/G NSs) for electrocatalytic water oxidation.: This article presents a study on the use of sodium hexanitrocobaltate(III) in a one-pot synthesis method for producing graphene-cobalt hydroxide composite nanosheets. These composites show significant potential for applications in electrocatalytic water oxidation, highlighting their importance in energy conversion and storage technologies (Mehmood R et al., 2018).

  • Nitrosation with Sodium Hexanitrocobaltate(III).: This research discusses the use of sodium hexanitrocobaltate(III) as a reagent for the nitrosation process. It provides valuable insights into the application of this compound in organic synthesis, demonstrating its effectiveness in introducing nitroso groups into organic molecules, which is critical for the development of pharmaceuticals and agrochemicals (Stefane B et al., 1997).

Analysis Note

Identity: passes test
Insoluble matter: ≤ 0.02 %
Chloride (Cl): ≤ 0.005 %
Fe (Iron): ≤ 0.002 %
Suitability for Potassium determination: passes test

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Ox. Sol. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service