Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.02498

Supelco

Chromotropic acid disodium salt dihydrate

for analysis ACS,Reag. Ph Eur

Synonym(s):

Chromotropic acid disodium salt dihydrate, 4,5-Dihydroxy-2,7-naphthalene disulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6Na2O8S2 · 2H2O
CAS Number:
Molecular Weight:
400.29
MDL number:
UNSPSC Code:
12352107
EC Index Number:
204-972-9
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

Assay

≥98.5% (acidimetric)

form

solid

composition

Water, 8.5-9.5% Karl Fischer

color

gray-white to brownish white

pH

3.6 (20 °C, 10 g/L in H2O)

mp

>300 °C

bulk density

780 kg/m3

storage temp.

2-30°C

InChI

1S/C10H8O8S2/c11-8-3-6(19(13,14)15)1-5-2-7(20(16,17)18)4-9(12)10(5)8/h1-4,11-12H,(H,13,14,15)(H,16,17,18)/p-2

InChI key

HLVXFWDLRHCZEI-UHFFFAOYSA-L

Linkage

Replaces: DX1460-1

Analysis Note

Appearance: greyish white to brownish white powder
Assay (acidimetric, calc. on anhydrous substance): ≥ 98.5 %
Identity (IR-spectrum): passes test
Appearance of solution (5 %; water): passes test
Water (according to Karl Fischer): 8.5 - 9.5 %
Sensitivity (of formaldehyde): passes test
Sensitivity (of nitrate): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service