Skip to Content
MilliporeSigma

1.08487

Urea

GR for analysis ACS,Reag. Ph Eur

Synonym(s):

Carbamide, Carbonyl diamide, Diaminomethanone, Carbonyldiamine, Carbamide, Carbonyldiamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
UNSPSC Code:
12352111
NACRES:
NA.21
EC Index Number:
200-315-5
MDL number:
Beilstein/REAXYS Number:
635724
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

agency

reag. Ph. Eur.

vapor pressure

<0.1 hPa ( 20 °C)

form

solid

potency

8471 mg/kg LD50, oral (Rat), 8200 mg/kg LD50, skin (Rat)

technique(s)

titration: suitable

pH

9 (20 °C, 100 g/L in H2O)

mp

132-135 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

bulk density

720‑760 kg/m3

storage temp.

15-25°C

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Application

Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Analysis Note

Assay (ex N): 99.0 - 100.5 %
Assay (ex N, calc. on dried substance): 99.0 - 101.5 %
Purity (DSC (differential scanning calorimetry)): ≥ 99.5 %
Identity (IR-spectrum): passes test
Appearance of solution (10 %; Wasser; colour): colourless
Appearance of solution (10 %; Wasser; clarity): clear
In water insoluble matter: ≤ 0.003 %
Acidity, Alkalinity: ≤ 0.0005 meq/g
Melting point (DSC): 132 - 135 °C
NH₄ (Ammonium): ≤ 0.0500 %
Chloride (Cl): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0004 %
Biuret: ≤ 0.05 %
Sulfated ash (600 °C): ≤ 0.005 %
Cu (Copper): ≤ 0.0001 %
Fe (Iron): ≤ 0.0001 %
Pb (Lead): ≤ 0.0002 %
Loss on Drying (105°C): ≤ 1.0 %
Corresponds to ACS, Reag. Ph Eur

Other Notes

Replaces: UX0065-1


Still not finding the right product?

Explore all of our products under Urea


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library