Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.01944

Supelco

Bromoform

for separation of minerals mixtures

Synonym(s):

Bromoform, Tribromomethane, Methane tribromide, Methyl tribromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHBr3
CAS Number:
Molecular Weight:
252.73
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-854-6
NACRES:
NB.21

vapor pressure

7.5 hPa ( 25 °C)

Quality Level

form

liquid

potency

933 mg/kg LD50, oral (Rat)

bp

149.5 °C/1013 mbar

mp

9.2 °C

solubility

3.2 g/L

density

2.89 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/CHBr3/c2-1(3)4/h1H

InChI key

DIKBFYAXUHHXCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Identity (IR-spectrum): passes test
Refractive index (n 20/D): 1.597 - 1.599
Density (d 20 °C/ 4 °C): 2.88 - 2.90
Evaporation residue: ≤ 0.1 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

106.3 °F - closed cup - (own results)

Flash Point(C)

41.3 °C - closed cup - (own results)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service