Skip to Content
MilliporeSigma
Bromoform amylene stabilized, analytical standard

Bromoform

Bromoform amylene stabilized, analytical standard
Synonym(s):
Tribromomethane, Bromoform
Empirical Formula (Hill Notation):
CHBr3
CAS Number:
Molecular Weight:
252.73
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique