Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

1.01770

Supelco

Petroleum benzine boiling range 100-140°C

(naphtha benzine) EMPLURA®

Synonym(s):

Petroleum ether, Petroleum spirit, Petroleum ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.21
bp:
107-137 °C/1013 hPa
vapor pressure:
30-40 hPa ( 20 °C)

vapor pressure

30-40 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

autoignition temp.

250 °C

expl. lim.

0.8-6.5 % (v/v)

dilution

(low-cost alternative, for basic applications)

impurities

≤0.0005% Free acid (as CH3COOH)
≤0.01% Water
≤0.05% Aromatics (as toluol)

evapn. residue

≤0.005%

kinematic viscosity

0.76 cSt(25 °C)

bp

107-137 °C/1013 hPa

transition temp

flash point <-10 °C

density

0.728 g/cm3 at 15 °C

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application



  • Ginkgo Biloba L. Exocarp Extracts: Utilizing petroleum benzine for solvent extraction, this research explores the anti-microbial potential of Ginkgo biloba L. exocarp extracts against methicillin-resistant Staphylococcus aureus, indicating its significant role in pharmaceutical applications (Wei et al., 2024).


  • Creating Mesopores in ZSM-5 Catalyst for Aromatics Production: This application of petroleum benzine in the synthesis of biodiesel-derived waste glycerol showcases innovative use in catalytic processes to enhance aromatic production, beneficial for both environmental and industrial chemistry (Kumar et al., 2024).


  • High-Speed Countercurrent Chromatography for Bioactive Neolignan Isomers: Demonstrating the use of petroleum benzine in chromatographic techniques, this method significantly aids in the isolation of complex organic compounds, crucial for advanced research in natural product chemistry (Xiao et al., 2024).


  • Hydrocarbon Ingestions and Poison Control: While indirectly related, the study uses data on hydrocarbon ingestions to emphasize the importance of safety and environmental impact assessments in the use of petroleum products, including benzine, within public health contexts (Palmer et al., 2023).


Analysis Note

Free acid (as CH₃COOH): ≤ 0.0005 %
Boiling range (lower value): ≥ 100.0 °C
Boiling range (upper value): ≤ 140.0 °C
Density (d 20 °C/ 4 °C): 0.720 - 0.733
Aromatics (as toluol): ≤ 0.05 %
Readily carbonizable substances: conforms
Evaporation residue: ≤ 0.005 %
Water: ≤ 0.01 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service