Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01676

Sigma-Aldrich

Histosec60 pastilles (without DMSO)

solidification point 58-60°C, embedding agent for histology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.21

Quality Level

color

light gray

transition temp

flash point >100 °C

bulk density

400 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Histosec 60 pastilles (without DMSO) - solidification point 58-60°C, are a ready-to-use embedding agent for histology used to embed specimens of animal origin. Histosec 60 pastilles (without DMSO) are a paraffin enriched with polymers that, for the protection of the user, contains no DMSO (dimethyl sulfoxide). The addition of polymers prevents sprinkling, air-filled slits between the paraffin crystals that can adversely affect the sectioning procedure. Thin sections can then be reliably prepared from the paraffin-embedded specimens (blocks) for further processing. Paraffin is a traditional embedding media for histology. Fixed and dehydrated specimens are embedded and blocked with paraffin with an appropriate solidification point. In order to achieve uniformous ribbon-sectioning quality, defined hardness and homogeneity of the processed material is necessary. This can be guaranteed through high-quality raw materials and defined melting points. The melting point of Paraffin is 56-58°C. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

Paraffin embedding medium. Solidification point 58-60°C

Analysis Note

Solidification point (rotating thermometer method)58 - 60 °CEffect of heating (at 65 °C/100 g)clear melt within max. 20 hoursXylol solubility (22°C-25°C/0,1g in 5ml)passes testCutting results (4 µm slices)conformsSolubility of slices in xylene<50 seconds

Legal Information

HISTOSEC is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service