Skip to Content
MilliporeSigma

1.15161

Histosec pastilles (without DMSO)

solidification point 56-58°C, embedding agent for histology

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.21
UNSPSC Code:
41116124
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

IVD

for in vitro diagnostic use

mp

56-58 °C

transition temp

flash point >100 °C

bulk density

400 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Histosec pastilles (without DMSO) - solidification point 56-58 °C embedding agent for histology is a ready-to-use embedding medium for histology of samples of human origin. It is a paraffin enriched with polymers, also available with the addition of DMSO (dimethyl sulfoxide) (Histosec pastilles (Product number 1.11609). DMSO increases the rate of penetration of paraffin and provides additional preservation of the tissue structures. Its melting range is between 56 - 58 °C and, within this range any specimen can be processed without fear of damage irrespective of the method used. In most clinical laboratories the paraffinization is performed with an automated embedding system and is performed directly after the histoprocessing. The Histosec pastilles (without DMSO) are well soluble with the Intermedia Xylene (isometic mixture) (Product number 1.08298) or xylene substitutes such as Neo-Clear(Product number 1.09843).
The paraffin is an IVD registered product and CE certified, thus can be used for clinical diagnostic purposes. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Legal Information

HISTOSEC is a trademark of Merck KGaA, Darmstadt, Germany
NEO-CLEAR is a trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_c

> 100 °C

flash_point_f

>212.0 °F



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library